Ethyl 3-(benzyloxy)-6-bromo-2-chlorobenzoate ≥97%
Supplier: AOBChem USA
|
Warning
|
37570-1G
37570-25G
37570-500MG
37570-5G
76941-872EA
290.69
USD
76941-872
76941-874
76941-876
76941-878
Ethyl 3-(benzyloxy)-6-bromo-2-chlorobenzoate ≥97%
Ethyl 3-(benzyloxy)-6-bromo-2-chlorobenzoate
Formula:
C₁₆H₁₄BrClO₃ MW: 369.638 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD32632240 CAS Number: 2404734-41-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...