6-Bromo-2-chloro-3-methoxyphenol ≥97% research grade

Supplier: AOBChem USA

Warning

37562-1G 37562-25G 37562-5G
76941-844EA 96.55 USD
76941-844 76941-846 76941-848
6-Bromo-2-chloro-3-methoxyphenol ≥97% research grade
6-Bromo-2-chloro-3-methoxyphenol
Formula: C₇H₆BrClO₂
Storage Temperature: Ambient
MDL Number: MFCD16618957
CAS Number: 1228957-06-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR