Methyl 4-fluoro-2,5-dimethylbenzoate ≥97%

Supplier: AOBChem USA

Warning

37515-1G 37515-500MG 37515-5G
76941-834EA 629.12 USD
76941-834 76941-836 76941-838
Methyl 4-fluoro-2,5-dimethylbenzoate ≥97%
Methyl 4-fluoro-2,5-dimethylbenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₁FO₂
MW: 182.192 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD28794299
CAS Number: 1806332-88-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR