tert-Butyl 6-chloro-4-methylnicotinate ≥97%

Supplier: AOBChem USA

Warning

37498-1G 37498-25G 37498-500MG 37498-5G
76941-818EA 290.69 USD
76941-818 76941-820 76941-822 76941-824
tert-Butyl 6-chloro-4-methylnicotinate ≥97%
tert-Butyl 6-chloro-4-methylnicotinate

Precaución: for Research Use Only

Formula: C₁₁H₁₄ClNO₂
MW: 227.687 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD32187962
CAS Number: 1649469-46-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR