tert-Butyl 2,4-bis(trifluoromethyl)benzoate ≥97%

Supplier: AOBChem USA

Warning

37309-25G 37309-5G
76941-790EA 453.01 USD
76941-790 76941-792
tert-Butyl 2,4-bis(trifluoromethyl)benzoate ≥97%
tert-Butyl 2,4-bis(trifluoromethyl)benzoate

Precaución: for Research Use Only

Formula: C₁₃H₁₂F₆O₂
MW: 314.224 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32902600
CAS Number: 2586126-78-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR