7-Iodo-[1,2,4]triazolo[4,3-a]pyridine ≥97%

Supplier: AOBChem USA

Warning

37267-1G 37267-25G 37267-5G
76941-780EA 98.67 USD
76941-780 76941-782 76941-784
7-Iodo-[1,2,4]triazolo[4,3-a]pyridine ≥97%
7-Iodo-[1,2,4]triazolo[4,3-a]pyridine

Precaución: for Research Use Only

Formula: C₆H₄IN₃
Storage Temperature: Refrigerator
MDL Number: MFCD27921215
CAS Number: 864866-44-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR