8-Bromo-6-methyl-[1,2,4]triazolo[4,3-a]pyridine ≥97%
Supplier: AOBChem USA
|
Warning
|
37195-1G
37195-25G
37195-5G
76941-738EA
71.09
USD
76941-738
76941-740
76941-742
8-Bromo-6-methyl-[1,2,4]triazolo[4,3-a]pyridine ≥97%
8-Bromo-6-methyl-[1,2,4]triazolo[4,3-a]pyridine
Precaución: for Research Use Only
Formula:
C₇H₆BrN₃ MW: 212.047 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD11847254 CAS Number: 1260769-17-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...