Ethyl 4-bromo-2-methoxybenzoate ≥95%

Supplier: AOBChem USA

Warning

37182-1G 37182-25G 37182-500MG 37182-5G
76941-714EA 274.78 USD
76941-714 76941-716 76941-718 76941-720
Ethyl 4-bromo-2-methoxybenzoate ≥95%
Ethyl 4-bromo-2-methoxybenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₁BrO₃
Storage Temperature: Refrigerator
MDL Number: MFCD11042782
CAS Number: 1214366-76-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR