tert-Butyl 3-fluoro-4-methylbenzoate ≥97%

Supplier: AOBChem USA

Warning

37179-25G 37179-5G
76941-702EA 418 USD
76941-702 76941-704
tert-Butyl 3-fluoro-4-methylbenzoate ≥97%
tert-Butyl 3-fluoro-4-methylbenzoate

Precaución: for Research Use Only

Formula: C₁₂H₁₅FO₂
MW: 210.245 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12913347
CAS Number: 866625-11-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR