Methyl 5-chloro-2-fluorobenzoate ≥97%

Supplier: AOBChem USA

Warning

37171-25G 37171-5G
76941-690EA 524.09 USD
76941-690 76941-692
Methyl 5-chloro-2-fluorobenzoate ≥97%
Methyl 5-chloro-2-fluorobenzoate

Precaución: for Research Use Only

Formula: C₈H₆ClFO₂
Storage Temperature: Refrigerator
MDL Number: MFCD06204184
CAS Number: 57381-36-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR