6-Bromo-2,8-dimethylimidazo[1,2-a]pyridine ≥95%

Supplier: AOBChem USA

Warning

37129-1G 37129-500MG 37129-5G
76941-680EA 445.58 USD
76941-680 76941-682 76941-684
6-Bromo-2,8-dimethylimidazo[1,2-a]pyridine ≥95%
6-Bromo-2,8-dimethylimidazo[1,2-a]pyridine

Precaución: for Research Use Only

Formula: C₉H₉BrN₂
MW: 225.085 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD14547282
CAS Number: 1216231-53-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR