Methyl 4'-methoxy-[1,1'-biphenyl]-4-carboxylate ≥97%

Supplier: AOBChem USA

Warning

37114-25G 37114-5G
76941-670EA 595.17 USD
76941-670 76941-672
Methyl 4'-methoxy-[1,1'-biphenyl]-4-carboxylate ≥97%
Methyl 4'-methoxy-[1,1'-biphenyl]-4-carboxylate

Precaución: for Research Use Only

Formula: C₁₅H₁₄O₃
MW: 242.27 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01122234
CAS Number: 729-17-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR