6-Chloro-8-fluoro[1,2,4]triazolo[4,3-a]pyridine ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 6-Chloro-8-fluoro-[1,2,4]triazolo[4,3-a]pyridine

36940-25G 36940-5G
76941-658EA 488.02 USD
76941-658 76941-660
6-Chloro-8-fluoro[1,2,4]triazolo[4,3-a]pyridine ≥97% research grade
6-Chloro-8-fluoro[1,2,4]triazolo[4,3-a]pyridine
Formula: C₆H₃ClFN₃
MW: 171.56 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09972195
CAS Number: 1020253-21-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR