Ethyl 2-fluoro-3-(trifluoromethyl)benzoate ≥97%

Supplier: AOBChem USA

Warning

36903-1G 36903-25G 36903-5G
76941-628EA 109.28 USD
76941-628 76941-630 76941-632
Ethyl 2-fluoro-3-(trifluoromethyl)benzoate ≥97%
Ethyl 2-fluoro-3-(trifluoromethyl)benzoate

Precaución: for Research Use Only

Formula: C₁₀H₈F₄O₂
MW: 236.163 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD06204409
CAS Number: 773134-92-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR