6-(2-Methoxyethoxy)nicotinic acid ≥97%

Supplier: AOBChem USA

Warning

36859-1G 36859-25G 36859-500MG 36859-5G
76941-612EA 245.07 USD
76941-612 76941-614 76941-616 76941-618
6-(2-Methoxyethoxy)nicotinic acid ≥97%
6-(2-Methoxyethoxy)nicotinic acid

Precaución: for Research Use Only

Formula: C₉H₁₁NO₄
Storage Temperature: Refrigerator
MDL Number: MFCD09812476
CAS Number: 894802-20-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR