6-Chloro[1,2,4]triazolo[4,3-a]pyridine ≥97%
Supplier: AOBChem USA
|
Warning
|
36825-1G
36825-25G
36825-500MG
36825-5G
76941-590EA
173.99
USD
76941-590
76941-592
76941-594
76941-596
6-Chloro[1,2,4]triazolo[4,3-a]pyridine ≥97%
6-Chloro[1,2,4]triazolo[4,3-a]pyridine
Formula:
C₆H₄ClN₃ Storage Temperature: Refrigerator |
MDL Number:
MFCD11100235 CAS Number: 1048913-70-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...