tert-Butyl 4-bromo-2-methoxybenzoate ≥97%

Supplier: AOBChem USA

Warning

36717-1G 36717-25G 36717-500MG 36717-5G
76941-570EA 260.99 USD
76941-570 76941-572 76941-574 76941-576
tert-Butyl 4-bromo-2-methoxybenzoate ≥97%
tert-Butyl 4-bromo-2-methoxybenzoate

Precaución: for Research Use Only

Formula: C₁₂H₁₅BrO₃
MW: 287.15 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08703421
CAS Number: 1260897-35-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR