7-Fluoro-3-iodo-1H-indazole ≥95%

Supplier: AOBChem USA

Warning

36695-1G 36695-25G 36695-5G
76941-560EA 62.6 USD
76941-560 76941-562 76941-564
7-Fluoro-3-iodo-1H-indazole ≥95%
7-Fluoro-3-iodo-1H-indazole
Formula: C₇H₄FIN₂
MW: 262.02 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD10696806
CAS Number: 944904-38-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR