Ethyl 5-bromo-2-methoxybenzoate ≥97%

Supplier: AOBChem USA

Warning

36551-1G 36551-25G 36551-500MG 36551-5G
76941-508EA 236.59 USD
76941-508 76941-510 76941-512 76941-514
Ethyl 5-bromo-2-methoxybenzoate ≥97%
Ethyl 5-bromo-2-methoxybenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₁BrO₃
Storage Temperature: Refrigerator
MDL Number: MFCD06204296
CAS Number: 773134-60-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR