Methyl 3-fluoro-5-(methylthio)benzoate ≥97%

Supplier: AOBChem USA

Warning

36364-1G 36364-25G 36364-500MG 36364-5G
76941-478EA 389.36 USD
76941-478 76941-480 76941-482 76941-484
Methyl 3-fluoro-5-(methylthio)benzoate ≥97%
Methyl 3-fluoro-5-(methylthio)benzoate

Precaución: for Research Use Only

Formula: C₉H₉FO₂S
MW: 200.23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32632002
CAS Number: 2379322-70-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR