tert-Butyl 2-(4-bromo-1H-pyrazol-1-yl)acetate ≥95% research grade

Supplier: AOBChem USA

Warning

36229-25G 36229-5G
76941-428EA 381.93 USD
76941-428 76941-430
tert-Butyl 2-(4-bromo-1H-pyrazol-1-yl)acetate ≥95% research grade
tert-Butyl 2-(4-bromo-1H-pyrazol-1-yl)acetate
Formula: C₉H₁₃BrN₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD13193178
CAS Number: 1199773-67-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR