Methyl 2,5-difluoro-4-methoxybenzoate ≥97%

Supplier: AOBChem USA

Warning

36184-1G 36184-25G 36184-500MG 36184-5G
76941-398EA 178.24 USD
76941-398 76941-400 76941-402 76941-404
Methyl 2,5-difluoro-4-methoxybenzoate ≥97%
Methyl 2,5-difluoro-4-methoxybenzoate

Precaución: for Research Use Only

Formula: C₉H₈F₂O₃
MW: 202.155 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18399129
CAS Number: 1261830-21-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR