4-Bromo-N-isopropyl-3-methylbenzamide ≥97%

Supplier: AOBChem USA

Warning

Synonyms: N-Isopropyl 4-bromo-3-methylbenzamide

35848-25G 35848-5G
76941-314EA 346.92 USD
76941-314 76941-316
4-Bromo-N-isopropyl-3-methylbenzamide ≥97%
4-Bromo-N-isopropyl-3-methylbenzamide

Precaución: for Research Use Only

Formula: C₁₁H₁₄BrNO
Storage Temperature: Ambient
MDL Number: MFCD09972122
CAS Number: 1020252-77-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR