5-Phenyl-1,3-oxazole ≥95% research grade

Supplier: AOBChem USA

Warning

35800-1G 35800-25G 35800-5G
76941-300EA 62.6 USD
76941-300 76941-302 76941-304
5-Phenyl-1,3-oxazole ≥95% research grade
5-Phenyl-1,3-oxazole
Formula: C9H7NO
Storage Temperature: Ambient
MDL Number: MFCD00067074
CAS Number: 1006-68-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR