2-Bromo-6-isopropylphenol ≥97%

Supplier: AOBChem USA

Warning

35521-1G 35521-25G 35521-500MG 35521-5G
76941-216EA 438.16 USD
76941-216 76941-218 76941-220 76941-222
2-Bromo-6-isopropylphenol ≥97%
2-Bromo-6-isopropylphenol

Precaución: for Research Use Only

Formula: C₉H₁₁BrO
Storage Temperature: Refrigerator
MDL Number: MFCD16997221
CAS Number: 129976-32-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR