Methyl 2,6-dichloro-4-methylbenzoate ≥97%

Supplier: AOBChem USA

Warning

35421-1G 35421-25G 35421-500MG 35421-5G
76941-192EA 124.13 USD
76941-192 76941-194 76941-196 76941-198
Methyl 2,6-dichloro-4-methylbenzoate ≥97%
Methyl 2,6-dichloro-4-methylbenzoate

Precaución: for Research Use Only

Formula: C₉H₈Cl₂O₂
Storage Temperature: Ambient
MDL Number: MFCD28142003
CAS Number: 1098620-09-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR