Methyl 2-bromo-6-fluoro-3-(trifluoromethyl)benzoate ≥97%

Supplier: AOBChem USA

Warning

35262-1G 35262-500MG 35262-5G
76941-156EA 635.48 USD
76941-156 76941-158 76941-160
Methyl 2-bromo-6-fluoro-3-(trifluoromethyl)benzoate ≥97%
Methyl 2-bromo-6-fluoro-3-(trifluoromethyl)benzoate
Formula: C₉H₅BrF₄O₂
MW: 301.032 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD31613607
CAS Number: 2090828-24-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR