Methyl 4-(ethoxymethyl)benzoate ≥97%

Supplier: AOBChem USA

Warning

35098-1G 35098-25G 35098-500MG 35098-5G
76941-126EA 177.18 USD
76941-126 76941-128 76941-130 76941-132
Methyl 4-(ethoxymethyl)benzoate ≥97%
Methyl 4-(ethoxymethyl)benzoate

Precaución: for Research Use Only

Formula: C₁₁H₁₄O₃
MW: 194.227 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11189997
CAS Number: 886531-74-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR