tert-Butyl (4-bromo-2-chlorophenyl)carbamate ≥97%

Supplier: AOBChem USA

Warning

35056-1G 35056-25G 35056-500MG 35056-5G
76941-100EA 206.88 USD
76941-100 76941-102 76941-104 76941-106
tert-Butyl (4-bromo-2-chlorophenyl)carbamate ≥97%
tert-Butyl (4-bromo-2-chlorophenyl)carbamate

Precaución: for Research Use Only

Formula: C₁₁H₁₃BrClNO₂
Storage Temperature: Refrigerator
MDL Number: MFCD08703148
CAS Number: 330794-09-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR