Ethyl 4-hydroxy-2-methylbenzoate ≥97%

Supplier: AOBChem USA

Warning

34939-1G 34939-25G 34939-5G
76941-054EA 88.06 USD
76941-054 76941-056 76941-058
Ethyl 4-hydroxy-2-methylbenzoate ≥97%
Ethyl 4-hydroxy-2-methylbenzoate
Formula: C₁₀H₁₂O₃
Storage Temperature: Ambient
MDL Number: MFCD12025348
CAS Number: 57081-00-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR