Ethyl 6-bromo-2-chloro-3-methylbenzoate ≥97%

Supplier: AOBChem USA

Warning

34918-1G 34918-25G 34918-500MG 34918-5G
76941-040EA 266.29 USD
76941-040 76941-042 76941-044 76941-046
Ethyl 6-bromo-2-chloro-3-methylbenzoate ≥97%
Ethyl 6-bromo-2-chloro-3-methylbenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₀BrClO₂
MW: 277.542 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD31805778
CAS Number: 1478503-61-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR