6-Bromo-2-fluoro-3-iodopyridine ≥95%

Supplier: AOBChem USA

Warning

34884-1G 34884-25G 34884-500MG 34884-5G
76941-024EA 292.81 USD
76941-024 76941-026 76941-028 76941-030
6-Bromo-2-fluoro-3-iodopyridine ≥95%
6-Bromo-2-fluoro-3-iodopyridine
Formula: C₅H₂BrFIN
Storage Temperature: Ambient
MDL Number: MFCD28139682
CAS Number: 1599474-54-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR