N-Cyclopropyl-5-bromo-3-methylpyridine-2-carboxamide ≥97%
Supplier: AOBChem USA
|
Warning
|
34753-1G
34753-25G
34753-500MG
34753-5G
76940-968EA
147.47
USD
76940-968
76940-970
76940-972
76940-974
N-Cyclopropyl-5-bromo-3-methylpyridine-2-carboxamide ≥97%
N-Cyclopropyl-5-bromo-3-methylpyridine-2-carboxamide
Precaución: for Research Use Only
Formula:
C₁₀H₁₁BrN₂O Storage Temperature: Ambient |
MDL Number:
MFCD31696104 CAS Number: 2230481-36-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...