Methyl 4-benzyloxy-2-chlorobenzoate ≥97%

Supplier: AOBChem USA

Warning

34709-1G 34709-500MG 34709-5G
76940-956EA 485.9 USD
76940-956 76940-958 76940-960
Methyl 4-benzyloxy-2-chlorobenzoate ≥97%
Methyl 4-benzyloxy-2-chlorobenzoate

Precaución: for Research Use Only

Formula: C₁₅H₁₃ClO₃
Storage Temperature: Ambient
MDL Number: MFCD31696201
CAS Number: 154605-91-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR