Ethyl 4-bromo-1,3-thiazole-2-carboxylate ≥97%

Supplier: AOBChem USA

Warning

Synonyms: Ethyl 4-bromothiazole-2-carboxylate

34688-1G 34688-25G 34688-5G
76940-942EA 64.72 USD
76940-942 76940-944 76940-946
Ethyl 4-bromo-1,3-thiazole-2-carboxylate ≥97%
Ethyl 4-bromo-1,3-thiazole-2-carboxylate

Precaución: for Research Use Only

Formula: C₆H₆BrNO₂S
Storage Temperature: Refrigerator
MDL Number: MFCD11044971
CAS Number: 959755-96-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR