6-Bromo-2-chloro-3-fluorobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

34643-1G 34643-500MG 34643-5G
76940-914EA 558.04 USD
76940-914 76940-916 76940-918
6-Bromo-2-chloro-3-fluorobenzaldehyde ≥97%
6-Bromo-2-chloro-3-fluorobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₃BrClFO
Storage Temperature: Refrigerator
MDL Number: MFCD28733870
CAS Number: 1806058-24-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR