Ethyl 2-bromo-6-chloro-3-methylbenzoate ≥95%

Supplier: AOBChem USA

Warning

34615-1G 34615-25G 34615-500MG 34615-5G
76940-898EA 183.54 USD
76940-898 76940-900 76940-902 76940-904
Ethyl 2-bromo-6-chloro-3-methylbenzoate ≥95%
Ethyl 2-bromo-6-chloro-3-methylbenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₀BrClO₂
Storage Temperature: Refrigerator
MDL Number: MFCD31619512
CAS Number: 1478503-69-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR