Ethyl 2,3-difluoro-4-methoxybenzoate ≥97%

Supplier: AOBChem USA

Warning

34610-1G 34610-25G 34610-5G
76940-886EA 107.16 USD
76940-886 76940-888 76940-890
Ethyl 2,3-difluoro-4-methoxybenzoate ≥97%
Ethyl 2,3-difluoro-4-methoxybenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₀F₂O₃
MW: 216.181 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11520399
CAS Number: 1806332-41-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR