Ethyl 6-(piperidin-1-yl)pyridine-3-carboxylate ≥97%
Supplier: AOBChem USA
|
Warning
|
34572-1G
34572-25G
34572-500MG
34572-5G
76940-826EA
374.5
USD
76940-826
76940-828
76940-830
76940-832
Ethyl 6-(piperidin-1-yl)pyridine-3-carboxylate ≥97%
Ethyl 6-(piperidin-1-yl)pyridine-3-carboxylate
Precaución: for Research Use Only
Formula:
C₁₃H₁₈N₂O₂ MW: 234.294 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD12403464 CAS Number: 1225065-46-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...