Methyl 2-Methyl-4-(Methylsulfanyl)Benzoate ≥97%
Supplier: AOBChem USA
|
Warning
|
34394-1G
34394-500MG
34394-5G
76940-766EA
683.22
USD
76940-766
76940-768
76940-770
Methyl 2-Methyl-4-(Methylsulfanyl)Benzoate ≥97%
Methyl 2-Methyl-4-(Methylsulfanyl)Benzoate
Precaución: for Research Use Only
Formula:
C₁₀H₁₂O₂S MW: 196.266 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD22489230 CAS Number: 1822631-90-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...