7-Bromo-6-fluoro-1H-indazole ≥97%

Supplier: AOBChem USA

Warning

34344-1G 34344-25G 34344-500MG 34344-5G
76940-752EA 439.22 USD
76940-752 76940-754 76940-756 76940-758
7-Bromo-6-fluoro-1H-indazole ≥97%
7-Bromo-6-fluoro-1H-indazole
Formula: C₇H₄BrFN₂
MW: 215.0225 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD23705437
CAS Number: 1427377-75-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR