Methyl 3,4,5-trichlorobenzoate ≥97%

Supplier: AOBChem USA

Warning

34342-1G 34342-25G 34342-500MG 34342-5G
76940-736EA 266.29 USD
76940-736 76940-738 76940-740 76940-742
Methyl 3,4,5-trichlorobenzoate ≥97%
Methyl 3,4,5-trichlorobenzoate

Precaución: for Research Use Only

Formula: C₈H₅Cl₃O₂
MW: 239.483 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14704255
CAS Number: 114821-66-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR