Methyl 3-bromo-2-fluoro-5-methylbenzoate ≥97%

Supplier: AOBChem USA

Warning

34301-1G 34301-25G 34301-500MG 34301-5G
76940-722EA 374.5 USD
76940-722 76940-724 76940-726 76940-728
Methyl 3-bromo-2-fluoro-5-methylbenzoate ≥97%
Methyl 3-bromo-2-fluoro-5-methylbenzoate

Precaución: for Research Use Only

Formula: C₉H₈BrFO₂
Storage Temperature: Ambient
MDL Number: MFCD12911494
CAS Number: 72518-21-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR