Methyl 3-chloro-2-fluoro-5-(trifluoromethyl)benzoate ≥97%

Supplier: AOBChem USA

Warning

34245-1G 34245-25G 34245-5G
76940-694EA 96.55 USD
76940-694 76940-696 76940-698
Methyl 3-chloro-2-fluoro-5-(trifluoromethyl)benzoate ≥97%
Methyl 3-chloro-2-fluoro-5-(trifluoromethyl)benzoate

Precaución: for Research Use Only

Formula: C₉H₅ClF₄O₂
MW: 256.581 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06203785
CAS Number: 773874-03-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR