Methyl 2-bromo-5-iodobenzoate ≥97%

Supplier: AOBChem USA

Warning

34129-25G 34129-5G
76940-654EA 381.93 USD
76940-654 76940-656
Methyl 2-bromo-5-iodobenzoate ≥97%
Methyl 2-bromo-5-iodobenzoate

Precaución: for Research Use Only

Formula: C₈H₆BrIO₂
Storage Temperature: Ambient
MDL Number: MFCD01851253
CAS Number: 717880-58-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR