Methyl 4-bromo-2,6-dichlorobenzoate ≥97%

Supplier: AOBChem USA

Warning

34120-1G 34120-25G 34120-5G
76940-630EA 62.6 USD
76940-630 76940-632 76940-634
Methyl 4-bromo-2,6-dichlorobenzoate ≥97%
Methyl 4-bromo-2,6-dichlorobenzoate
Formula: C₈H₅BrCl₂O₂
MW: 283.93 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD28037387
CAS Number: 232275-53-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR