Ethyl 3-bromo-5-chloropicolinate ≥97%
Supplier: AOBChem USA
|
Warning
|
34093-1G
34093-25G
34093-5G
76940-620EA
88.06
USD
76940-620
76940-622
76940-624
Ethyl 3-bromo-5-chloropicolinate ≥97%
Ethyl 3-bromo-5-chloropicolinate
Precaución: for Research Use Only
Formula:
C₈H₇BrClNO₂ MW: 264.504 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD14698311 CAS Number: 1214357-95-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...