6-Bromo-2-chloro-3-fluorophenol ≥95%

Supplier: AOBChem USA

Warning

34022-1G 34022-25G 34022-500MG 34022-5G
76940-606EA 260.99 USD
76940-606 76940-608 76940-610 76940-612
6-Bromo-2-chloro-3-fluorophenol ≥95%
6-Bromo-2-chloro-3-fluorophenol
Formula: C₆H₃BrClFO
Storage Temperature: Refrigerator
MDL Number: MFCD28734041
CAS Number: 1805518-71-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR