Methyl-5-bromobenzo[b]thiophene-2-carboxylate ≥97%

Supplier: AOBChem USA

Warning

Synonyms: Methyl 5-bromobenzo[b]thiophene-2-carboxylate, Methyl 5-bromo-1-benzothiophene-2-carboxylate

33998-25G 33998-5G
76940-594EA 418 USD
76940-594 76940-596
Methyl-5-bromobenzo[b]thiophene-2-carboxylate ≥97%
Methyl 5-bromobenzo[b]thiophene-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₇BrO₂S
MW: 271.1300 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05662034
CAS Number: 7312-11-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR