Methyl-2,4,6-trifluorobenzoate ≥97%

Supplier: AOBChem USA

Warning

Synonyms: Methyl 2,4,6-trifluorobenzoate

33770-25G 33770-5G
76940-584EA 325.7 USD
76940-584 76940-586
Methyl-2,4,6-trifluorobenzoate ≥97%
Methyl 2,4,6-trifluorobenzoate

Precaución: for Research Use Only

Formula: C₈H₅F₃O₂
Storage Temperature: Ambient
MDL Number: MFCD01090991
CAS Number: 79538-28-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR